Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-C2-amido-C2-COOH | 2353496-84-9 | 98.5% | 25 MG
SDP

Catalog No. 5000341751
Encompass_Preferred

Thalidomide-C2-amido-C2-COOH incorporates a CRBN ligand for the E3 ubiquitin ligase and a linker. This compound is suitable for designing PROTAC CDK2/9 Degrader-1.

  • Incorporates a CRBN ligand for the E3 ubiquitin ligase and a linker
  • Targets cereblon
  • Has a molecular weight of 416.38
  • Has a molecular formula of C19H20N4O7
  • Appears as a solid with a light yellow to yellow color
  • Purity of 98.45%
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic)
  • Powder storage: -20°C for 3 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-41751 Supplier Medchemexpress LLC Supplier No. 25MGHY130713
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-C2-amido-C2-COOH incorporates a CRBN ligand for the E3 ubiquitin ligase and a linker. This compound is suitable for designing PROTAC CDK2/9 Degrader-1.

  • Incorporates a CRBN ligand for the E3 ubiquitin ligase and a linker
  • Targets cereblon
  • Has a molecular weight of 416.38
  • Has a molecular formula of C19H20N4O7
  • Appears as a solid with a light yellow to yellow color
  • Purity of 98.45%
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic)
  • Powder storage: -20°C for 3 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.