Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-C2-amido-C2-COOH | 2353496-84-9 | 98.5% | 416.38 g/mol | C19H20N4O7 | 5 MG
SDP

Catalog No. 5000262135
Encompass_Preferred

Thalidomide-C2-amido-C2-COOH is an E3 ligase ligand-linker conjugate that incorporates a cereblon-binding thalidomide-derived moiety and a carboxylic acid linker. It is intended for use as a research reagent in PROTAC design and related chemical biology studies; not for human or clinical use.

  • CAS number: 2353496-84-9.
  • Molecular formula: C19H20N4O7.
  • Molecular weight: 416.38 g/mol.
  • Purity: 98.45% (typical by supplier).
  • Solubility: DMSO up to 100 mg/mL (may require sonication).
  • Storage: powder at -20°C; stock solutions at -80°C for long term.
  • Intended use: CRBN ligand for PROTAC design and E3 ligase studies.
  • Available in small mg quantities for research use.

Catalog No. 50-002-62135 Supplier Medchemexpress LLC Supplier No. HY1307135MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-C2-amido-C2-COOH is an E3 ligase ligand-linker conjugate that incorporates a cereblon-binding thalidomide-derived moiety and a carboxylic acid linker. It is intended for use as a research reagent in PROTAC design and related chemical biology studies; not for human or clinical use.

  • CAS number: 2353496-84-9.
  • Molecular formula: C19H20N4O7.
  • Molecular weight: 416.38 g/mol.
  • Purity: 98.45% (typical by supplier).
  • Solubility: DMSO up to 100 mg/mL (may require sonication).
  • Storage: powder at -20°C; stock solutions at -80°C for long term.
  • Intended use: CRBN ligand for PROTAC design and E3 ligase studies.
  • Available in small mg quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.