Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-C2-amido-C2-COOH | 2353496-84-9 | 98.5% | C19H20N4O7 | 10MG
SDP

Catalog No. 5000261166
Encompass_Preferred

Thalidomide-C2-amido-C2-COOH is a thalidomide-derived cereblon (CRBN) ligand-linker conjugate for research use. Supplied as a powder, it is intended as a building block for design and synthesis of PROTAC degraders and other molecules that recruit the CRBN E3 ubiquitin ligase.

  • Incorporates a cereblon-binding thalidomide moiety for E3 ligase recruitment.
  • Contains a short C2-amido-C2-COOH linker for conjugation to target ligands.
  • High purity (98.5%) suitable for synthetic and analytical work.
  • Molecular formula and weight provided to support design calculations.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Powder form allows long-term frozen storage for stability.

Catalog No. 50-002-61166 Supplier Medchemexpress LLC Supplier No. HY13071310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-C2-amido-C2-COOH is a thalidomide-derived cereblon (CRBN) ligand-linker conjugate for research use. Supplied as a powder, it is intended as a building block for design and synthesis of PROTAC degraders and other molecules that recruit the CRBN E3 ubiquitin ligase.

  • Incorporates a cereblon-binding thalidomide moiety for E3 ligase recruitment.
  • Contains a short C2-amido-C2-COOH linker for conjugation to target ligands.
  • High purity (98.5%) suitable for synthetic and analytical work.
  • Molecular formula and weight provided to support design calculations.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Powder form allows long-term frozen storage for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.