Learn More
Medchemexpress LLC Thalidomide-CH2NH2 (hydrochloride) | 444287-40-5 | 98.1% | 323.73 | 5 MG
SDP
Catalog No. 5000404181
Encompass_Preferred
Thalidomide-CH2NH2 hydrochloride is an analogue of Thalidomide that includes a primary amine, a versatile functional group capable of participating in various reactions. This product is intended for research use only and is not sold to patients.
  • Analogue
  • Features a primary amine, allowing for versatile reactions
Catalog No. 50-004-04181 Supplier Medchemexpress LLC Supplier No. HY442495MG
Thalidomide-CH2NH2 hydrochloride is an analogue of Thalidomide that includes a primary amine, a versatile functional group capable of participating in various reactions. This product is intended for research use only and is not sold to patients.
  • Analogue
  • Features a primary amine, allowing for versatile reactions