Learn More
Medchemexpress LLC Thalidomide-CH2NH2 hydrochloride | 444287-40-5 | MFCD32183715 | 98.0% | 323.73 g/mol | C14H14ClN3O4 | 10 MG
SDP
Catalog No. 5000299067
Encompass_Preferred
Thalidomide-CH2NH2 hydrochloride is a thalidomide analogue supplied as the hydrochloride salt for research use. It features a primary aminomethyl substituent that enables further functionalization and utility as a ligand-like scaffold in biochemical studies. The compound is provided as a high-purity solid suitable for synthetic chemistry and assay development.
  • Contains a primary aminomethyl group for synthetic functionalization.
  • Supplied as the hydrochloride salt for improved stability and handling.
  • High purity suitable for research and assay development.
  • Serves as a ligand scaffold for E3 ligase-related studies.
  • Available in small-scale quantities for discovery workflows.
Catalog No. 50-002-99067 Supplier Medchemexpress LLC Supplier No. HY4424910MG
Thalidomide-CH2NH2 hydrochloride is a thalidomide analogue supplied as the hydrochloride salt for research use. It features a primary aminomethyl substituent that enables further functionalization and utility as a ligand-like scaffold in biochemical studies. The compound is provided as a high-purity solid suitable for synthetic chemistry and assay development.
  • Contains a primary aminomethyl group for synthetic functionalization.
  • Supplied as the hydrochloride salt for improved stability and handling.
  • High purity suitable for research and assay development.
  • Serves as a ligand scaffold for E3 ligase-related studies.
  • Available in small-scale quantities for discovery workflows.