Learn More
Cayman Chemical -ThalIdomIde-d4 1mg
SDP
Supplier:  Cayman Chemical 294501 MG
Encompass_Preferred
An internal standard for the quantification of (±)-thalidomide by GC- or LC-MS
Catalog No. 50-297-8876