Learn More
Cayman Chemical (±)-Thalidomide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1219177-18-0, 5 mg
SDP
Catalog No. 502978984
Encompass_Preferred
An internal standard for the quantification of (±)-thalidomide by GC- or LC-MS
Catalog No. 50-297-8984 Supplier Cayman Chemical Supplier No. 294505 MG
An internal standard for the quantification of (±)-thalidomide by GC- or LC-MS