Learn More
Medchemexpress LLC HY-41547 10mg Medchemexpress, Thalidomide fluoride CAS:835616-60-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY4154710MG
Encompass_Preferred
Medchemexpress, HY-41547 10mg Thalidomide fluoride CAS:835616-60-9 Thalidomide fluoride (Cereblon ligand 4) is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide fluoride (Cereblon ligand 4) can be connected to the ligand for IRAK4 protein by a linker to form PROTAC IRAK4 degrader-1 (HY-129966). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0053
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.