Learn More
Medchemexpress LLC Thalidomide-iodo | 2509137-01-1 | MFCD31735039 | 98.0% | 384.13 | C13H9IN2O4 | 100 MG
SDP
Catalog No. 5000285133
Encompass_Preferred
Thalidomide-iodo is an iodinated derivative of thalidomide supplied as a research reagent. It acts as an E3 ligase activator and is suitable as a synthetic building block for further derivatization via substitution of the iodine. Typical specifications include CAS 2509137-01-1, molecular formula C13H9IN2O4, molecular weight 384.13, and reported purity of 98.0%.
  • Iodinated E3 ligase activator for probe and degrader research.
  • Synthetic building block amenable to substitution of iodine.
  • High purity suitable for analytical and synthetic workflows.
  • Available in multiple pack sizes for small-scale research.
  • Intended for laboratory research use only.
Catalog No. 50-002-85133 Supplier Medchemexpress LLC Supplier No. HYW586331100MG
Thalidomide-iodo is an iodinated derivative of thalidomide supplied as a research reagent. It acts as an E3 ligase activator and is suitable as a synthetic building block for further derivatization via substitution of the iodine. Typical specifications include CAS 2509137-01-1, molecular formula C13H9IN2O4, molecular weight 384.13, and reported purity of 98.0%.
  • Iodinated E3 ligase activator for probe and degrader research.
  • Synthetic building block amenable to substitution of iodine.
  • High purity suitable for analytical and synthetic workflows.
  • Available in multiple pack sizes for small-scale research.
  • Intended for laboratory research use only.