Learn More
Medchemexpress LLC Thalidomide-iodo | 2509137-01-1 | MFCD31735039 | >98.0% | 384.13 g/mol | C13H9IN2O4 | 1 G
SDP
Catalog No. 5000283966
Encompass_Preferred
Thalidomide-iodo is an iodinated thalidomide derivative that acts as an E3 ligase activator and ligand for cereblon, used in targeted protein degradation and PROTAC research. The iodine substituent enables further derivatization through cross-coupling or substitution, supporting synthesis of linker conjugates and chemical probes. Supplied as a high-purity research chemical.
Catalog No. 50-002-83966 Supplier Medchemexpress LLC Supplier No. HYW5863311G
Thalidomide-iodo is an iodinated thalidomide derivative that acts as an E3 ligase activator and ligand for cereblon, used in targeted protein degradation and PROTAC research. The iodine substituent enables further derivatization through cross-coupling or substitution, supporting synthesis of linker conjugates and chemical probes. Supplied as a high-purity research chemical.