Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-iodo | 2509137-01-1 | MFCD31735039 | 98.0% | 384.13 g/mol | C13H9IN2O4 | 250 MG
SDP

Catalog No. 5000284871
Encompass_Preferred

Thalidomide-iodo is an iodinated derivative of thalidomide that functions as an E3 ligase activator for chemical biology and medicinal chemistry research. It is supplied as a solid research reagent and can be used as a synthetic building block for further derivatization via substitution of the iodine atom.

  • Iodinated thalidomide derivative suitable for E3 ligase activation studies.
  • Useful as a synthetic building block for iodine substitution and derivatization.
  • High purity (≥98.0%) appropriate for research applications.
  • Molecular weight 384.13 g/mol; formula C13H9IN2O4.
  • CAS number 2509137-01-1 for unambiguous identification.

Catalog No. 50-002-84871 Supplier Medchemexpress LLC Supplier No. HYW586331250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-iodo is an iodinated derivative of thalidomide that functions as an E3 ligase activator for chemical biology and medicinal chemistry research. It is supplied as a solid research reagent and can be used as a synthetic building block for further derivatization via substitution of the iodine atom.

  • Iodinated thalidomide derivative suitable for E3 ligase activation studies.
  • Useful as a synthetic building block for iodine substitution and derivatization.
  • High purity (≥98.0%) appropriate for research applications.
  • Molecular weight 384.13 g/mol; formula C13H9IN2O4.
  • CAS number 2509137-01-1 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.