Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thalidomide, MP Biomedicals™

Catalog No. ICN15875383 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15875383 250 mg
ICN15875380 100 mg
ICN15875350 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15875383 Supplier MP Biomedicals, Inc Supplier No. 0215875383
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50-35-1
Molecular Formula C13H10N2O4
Molecular Weight (g/mol) 258.23
MDL Number MFCD00153873
InChI Key UEJJHQNACJXSKW-UHFFFAOYNA-N
Synonym thalidomide, thalomid, +/--thalidomide, sedoval, 2-2,6-dioxopiperidin-3-yl isoindoline-1,3-dione, corronarobetin, psycholiquid, psychotablets, theophilcholine, algosediv
PubChem CID 5426
ChEBI CHEBI:74947
IUPAC Name 2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1N(C2CCC(=O)NC2=O)C(=O)C2=CC=CC=C12

Specifications

Color White
Quantity 250 mg
Physical Form Solid
Chemical Name or Material Thalidomide
EINECSNumber 200-031-1
storageNote1 Protect from light
Recommended Storage Store at room temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.