Learn More
Medchemexpress LLC Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride | 2983037-00-7 | 98.0% | 586.03 | 100 MG
SDP
Catalog No. 5000350073
Encompass_Preferred
Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. For research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Utilizes a linker in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-50073 Supplier Medchemexpress LLC Supplier No. HY138857A100MG
Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. For research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Utilizes a linker in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins