Learn More
Medchemexpress LLC Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride | 2983037-00-7 | >98.0% | 586.03 | 25 MG
SDP
Catalog No. 5000348417
Encompass_Preferred
Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It integrates a thalidomide-based cereblon ligand with a linker, making it suitable for applications in PROTAC technology. This compound is intended for research purposes only and is not for sale to patients.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide-based cereblon ligand
  • Utilizes a linker for PROTAC technology applications
  • Suitable for research use
Catalog No. 50-003-48417 Supplier Medchemexpress LLC Supplier No. 25MGHY138857A
Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It integrates a thalidomide-based cereblon ligand with a linker, making it suitable for applications in PROTAC technology. This compound is intended for research purposes only and is not for sale to patients.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide-based cereblon ligand
  • Utilizes a linker for PROTAC technology applications
  • Suitable for research use