Learn More
Medchemexpress LLC Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride | 2983037-00-7 | 98.0% | 586.03 | C25H36ClN5O9 | 5 MG
SDP
Catalog No. 5000223415
Encompass_Preferred
Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a PEG4-C2 spacer. Supplied as the hydrochloride salt in solid form and reported at high purity, the compound is intended as a building block for PROTAC design and other CRBN-recruiting chemical biology applications.
  • E3 ligase ligand-linker conjugate for PROTAC construction.
  • Includes a thalidomide-based cereblon ligand to recruit CRBN proteins.
  • PEG4-C2 linker with terminal amine for facile conjugation.
  • Hydrochloride salt form to aid handling and solubility.
  • High purity suitable for synthetic and research workflows.
Catalog No. 50-002-23415 Supplier Medchemexpress LLC Supplier No. HY138857A5MG
Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a PEG4-C2 spacer. Supplied as the hydrochloride salt in solid form and reported at high purity, the compound is intended as a building block for PROTAC design and other CRBN-recruiting chemical biology applications.
  • E3 ligase ligand-linker conjugate for PROTAC construction.
  • Includes a thalidomide-based cereblon ligand to recruit CRBN proteins.
  • PEG4-C2 linker with terminal amine for facile conjugation.
  • Hydrochloride salt form to aid handling and solubility.
  • High purity suitable for synthetic and research workflows.