Learn More
Medchemexpress LLC Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride | 2983037-00-7 | 98.01% | 586.03 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY138857A50MG
Encompass_Preferred
Thalidomide-NH-amido-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand. It is used in PROTAC technology, which exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Incorporates Thalidomide based cereblon ligand
  • Used in PROTAC technology
  • Targets E3 ubiquitin ligase
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-52388