Learn More
Medchemexpress LLC Thalidomide-NH-C10-COOH | 2428400-33-1 | 97.5% | 250 MG
SDP
Catalog No. 5000342670
Encompass_Preferred
Thalidomide-NH-C10-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide based CRBN ligand and a linker used in PROTAC technology. With a molecular weight of 457.52 and formula C24H31N3O6, this compound appears as a light yellow to yellow solid. It is intended for research use only.
  • Targets cereblon.
  • Recommended storage for powder is -20°C for 3 years or 4°C for 2 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Part of PROTAC technology.
Catalog No. 50-003-42670 Supplier Medchemexpress LLC Supplier No. 250MGHY130716
Thalidomide-NH-C10-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide based CRBN ligand and a linker used in PROTAC technology. With a molecular weight of 457.52 and formula C24H31N3O6, this compound appears as a light yellow to yellow solid. It is intended for research use only.
  • Targets cereblon.
  • Recommended storage for powder is -20°C for 3 years or 4°C for 2 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Part of PROTAC technology.