Learn More
Medchemexpress LLC Thalidomide-NH-C10-NH2 hydrochloride | 2460022-53-9 | 464.99 | C23H33ClN4O4 | 10 MG
SDP
Catalog No. 5000288145
Encompass_Preferred
Thalidomide-NH-C10-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as a hydrochloride salt in solid form for research use. It is intended for incorporation into PROTACs and other E3 ligase recruiting conjugates for chemical biology and medicinal chemistry studies.
  • Thalidomide-derived cereblon ligand for E3 ligase recruitment.
  • Suitable for PROTAC construction and linker conjugation.
  • Provided as a solid hydrochloride salt for stability and handling.
  • Available in small milligram packages appropriate for synthesis and assays.
  • Store sealed, away from moisture and light; follow supplier storage guidance.
Catalog No. 50-002-88145 Supplier Medchemexpress LLC Supplier No. HY15616010MG
Thalidomide-NH-C10-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as a hydrochloride salt in solid form for research use. It is intended for incorporation into PROTACs and other E3 ligase recruiting conjugates for chemical biology and medicinal chemistry studies.
  • Thalidomide-derived cereblon ligand for E3 ligase recruitment.
  • Suitable for PROTAC construction and linker conjugation.
  • Provided as a solid hydrochloride salt for stability and handling.
  • Available in small milligram packages appropriate for synthesis and assays.
  • Store sealed, away from moisture and light; follow supplier storage guidance.