Learn More
Selleck Chemical LLC Thalidomide-NH-C4-NH-Boc
SDP
Supplier:  Selleck Chemical LLC S63975MG
Encompass_Preferred
Thalidomide-NH-C4-NH-Boc is a novel potent and selective class of Bromodomain-containing protein 4 (BRD4) and Bromodomain-containing protein 2 (BRD2) degrader for the development of therapeutics to treat cancers
Catalog No. 50-000-03994