Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-nh-c4-nh-boc | 2093388-52-2 | 99.6% | 444.48 g/mol | C22H28N4O6 | 5 MG
SDP

Catalog No. 5000263021
Encompass_Preferred

Thalidomide-NH-C4-NH-Boc is a thalidomide-derived E3 ligase ligand-linker conjugate developed for PROTAC research. It combines a cereblon-binding thalidomide moiety with a Boc-protected four-carbon linker to facilitate targeted protein degradation studies. For research use only.

  • High purity: 99.64% by analytical HPLC.
  • Appropriate for PROTAC development as a cereblon ligand-linker conjugate.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: store at 4°C and protect from light; in solvent, -80°C (6 months) or -20°C (1 month).
  • Available in small mg quantities suitable for medicinal chemistry applications.

Catalog No. 50-002-63021 Supplier Medchemexpress LLC Supplier No. HY1306395MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-NH-C4-NH-Boc is a thalidomide-derived E3 ligase ligand-linker conjugate developed for PROTAC research. It combines a cereblon-binding thalidomide moiety with a Boc-protected four-carbon linker to facilitate targeted protein degradation studies. For research use only.

  • High purity: 99.64% by analytical HPLC.
  • Appropriate for PROTAC development as a cereblon ligand-linker conjugate.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: store at 4°C and protect from light; in solvent, -80°C (6 months) or -20°C (1 month).
  • Available in small mg quantities suitable for medicinal chemistry applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.