Learn More
Medchemexpress LLC Thalidomide-NH-C5-NH2 hydrochloride | 2375194-03-7 | 99.86% | 394.85 | 50 MG
SDP
Catalog No. 5000348337
Encompass_Preferred
Thalidomide-NH-C5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. For research use only.
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Contains two different ligands connected by a linker
Catalog No. 50-003-48337 Supplier Medchemexpress LLC Supplier No. HY13623750MG
Thalidomide-NH-C5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. For research use only.
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Contains two different ligands connected by a linker