Learn More
Medchemexpress LLC Thalidomide-NH-C5-NH2 hydrochloride | 2375194-03-7 | 99.9% | 394.85 | 100 MG
SDP
Catalog No. 5000348335
Encompass_Preferred
Thalidomide-NH-C5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the thalidomide-based cereblon ligand and a linker used in PROTAC technology. This conjugate exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide-based cereblon ligand
  • Used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-48335 Supplier Medchemexpress LLC Supplier No. HY136237100MG
Thalidomide-NH-C5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the thalidomide-based cereblon ligand and a linker used in PROTAC technology. This conjugate exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide-based cereblon ligand
  • Used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins