Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-NH-C5-NH2 hydrochloride | 2375194-03-7 | 99.9% | 394.85 | 250 MG
SDP

Catalog No. 5000348854
Encompass_Preferred

Thalidomide-NH-C5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. It is for research use only and not sold to patients.

  • Targets cereblon.
  • Utilizes PROTAC technology for selective degradation of target proteins.
  • Soluble in DMSO at 250 mg/mL (633.15 mM) with ultrasonication.
  • In vivo dissolution protocols available for clear solutions.
  • Ships at room temperature in continental US.
  • Solid storage at -20°C, sealed, away from moisture and light.
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-003-48854 Supplier Medchemexpress LLC Supplier No. 250MGHY136237
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-NH-C5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. It is for research use only and not sold to patients.

  • Targets cereblon.
  • Utilizes PROTAC technology for selective degradation of target proteins.
  • Soluble in DMSO at 250 mg/mL (633.15 mM) with ultrasonication.
  • In vivo dissolution protocols available for clear solutions.
  • Ships at room temperature in continental US.
  • Solid storage at -20°C, sealed, away from moisture and light.
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.