Learn More
Medchemexpress LLC Thalidomide-nh-c6-nh-boc | 2093536-13-9 | 96.1% | 100 MG
SDP
Catalog No. 5000338937
Encompass_Preferred
Thalidomide-NH-C6-NH-Boc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used for MI-389 (compound 22) synthesis. MI-389 is a potent phthalimide PROTAC degrader based on the multi-targeted receptor tyrosine kinase inhibitor sunitinib (HY-10255A).
  • Decreases cell growth with an EC50 value of 21.3 nM (comparable to sunitinib)
  • Leads to GSPT1 destabilization in a dose-dependent manner, showing complete GSPT1 depletion at 100 nM.
Catalog No. 50-003-38937 Supplier Medchemexpress LLC Supplier No. HY130854100MG
Thalidomide-NH-C6-NH-Boc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used for MI-389 (compound 22) synthesis. MI-389 is a potent phthalimide PROTAC degrader based on the multi-targeted receptor tyrosine kinase inhibitor sunitinib (HY-10255A).
  • Decreases cell growth with an EC50 value of 21.3 nM (comparable to sunitinib)
  • Leads to GSPT1 destabilization in a dose-dependent manner, showing complete GSPT1 depletion at 100 nM.