Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-NH-C6-NH2 hydrochloride | 2375194-37-7 | 99.6% | 100 MG
SDP

Catalog No. 5000340592
Encompass_Preferred

Thalidomide-NH-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates a Thalidomide-based cereblon ligand and a linker commonly used in PROTAC technology. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only and is not sold to patients.

  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a thalidomide-based cereblon ligand
  • Features a linker used in PROTAC technology
  • Designed to exploit the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight: 408.88
  • Chemical formula: C19H25ClN4O4
  • Appearance: Light yellow to yellow solid
  • Recommended storage: -20°C in sealed container, away from moisture and light
  • Long-term solvent storage: 6 months at -80°C or 1 month at -20°C

Catalog No. 50-003-40592 Supplier Medchemexpress LLC Supplier No. HY129704B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-NH-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates a Thalidomide-based cereblon ligand and a linker commonly used in PROTAC technology. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only and is not sold to patients.

  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a thalidomide-based cereblon ligand
  • Features a linker used in PROTAC technology
  • Designed to exploit the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight: 408.88
  • Chemical formula: C19H25ClN4O4
  • Appearance: Light yellow to yellow solid
  • Recommended storage: -20°C in sealed container, away from moisture and light
  • Long-term solvent storage: 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.