Learn More
Medchemexpress LLC Thalidomide-NH-C6-NH2 (hydrochloride) | 2375194-37-7 | 99.6% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY129704B
Encompass_Preferred
Thalidomide-NH-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Used in PROTAC technology
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-41535