Learn More
Medchemexpress LLC Thalidomide-NH-C6-NH2 hydrochloride | 2375194-37-7 | 99.6% | 50 MG
SDP
Catalog No. 5000341118
Encompass_Preferred
Thalidomide-NH-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates cereblon ligand and a linker used in PROTAC technology
  • For research use only
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-41118 Supplier Medchemexpress LLC Supplier No. HY129704B50MG
Thalidomide-NH-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates cereblon ligand and a linker used in PROTAC technology
  • For research use only
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins