Learn More
Medchemexpress LLC Thalidomide-NH-C6-NH2 TFA | 2093386-51-5 | 99.8% | 1 G
SDP
Catalog No. 5000341179
Encompass_Preferred
Thalidomide-NH-C6-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate, incorporating a Thalidomide-based cereblon ligand and a linker used in PROTAC technology. This compound is characterized by its high purity of 99.8%.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Features a linker used in PROTAC technology
  • Designed for use in PROTACs that exploit the intracellular ubiquitin-proteasome system
Catalog No. 50-003-41179 Supplier Medchemexpress LLC Supplier No. HY129704B1G
Thalidomide-NH-C6-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate, incorporating a Thalidomide-based cereblon ligand and a linker used in PROTAC technology. This compound is characterized by its high purity of 99.8%.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Features a linker used in PROTAC technology
  • Designed for use in PROTACs that exploit the intracellular ubiquitin-proteasome system