Learn More
Medchemexpress LLC Thalidomide-NH-C6-NH2 TFA | 2093386-51-5 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY129704A100MG
Encompass_Preferred
Thalidomide-NH-C6-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. This product is for research use only and not intended for sale to patients.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Utilizes linker used in PROTAC technology
  • For research use only
Catalog No. 50-003-38235