Learn More
Medchemexpress LLC Thalidomide-NH-C6-NH2 TFA | 2093386-51-5 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY129704A50MG
Encompass_Preferred
Thalidomide-NH-C6-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a Thalidomide-based cereblon ligand
  • Uses a linker in PROTAC technology
  • Targets Cereblon
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-39079