Learn More
Medchemexpress LLC Thalidomide-NH-C6-NH2 TFA | 2093386-51-5 | 99.8% | 500 MG
Supplier: Medchemexpress LLC HY129704A500MG
Thalidomide-NH-C6-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a linker used in PROTAC technology. This product is for research use only and not sold to patients.
- Synthesized E3 ligase ligand-linker conjugate
- Incorporates a Thalidomide-based cereblon ligand
- Used in PROTAC technology
- For research use only
- Related to autophagy and apoptosis