Learn More
Medchemexpress LLC Thalidomide-NH-C9-NH2 hydrochloride | 2305936-77-8 | 99.9% | 450.96 g/mol | C22H31ClN4O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15615910MG
Encompass_Preferred
Thalidomide-NH-C9-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as a hydrochloride salt for use in proteolysis-targeting chimera (PROTAC) construction and related chemical biology research. Provided as a light yellow solid with high purity and defined storage stability, it is intended for laboratory research use only.
  • High purity (99.85%) suitable for research applications.
  • Hydrochloride salt form to aid handling and solubility considerations.
  • Available in small research pack sizes, including 10 mg, for synthesis workflows.
  • Designed to act as a CRBN-binding E3 ligase ligand in PROTAC constructs.
  • Recommended storage: -20°C sealed; in solvent: -80°C up to 6 months or -20°C up to 1 month.
Catalog No. 50-002-87765