Learn More
Medchemexpress LLC Thalidomide-NH-CBP/p300 ligand 2 | 2484739-21-9 | 98.55% | 922.03 | 50 MG
SDP
Catalog No. 5000351578
Encompass_Preferred
Thalidomide-NH-CBP/p300 ligand 2 (P-007) is a PROTAC-based CBP and p300 degrader. It is for research use only and not sold to patients. This compound is extracted from patent WO2020173440 and targets Cereblon.
  • PROTAC-based degrader
  • Targets CBP and p300
  • For research use only
Catalog No. 50-003-51578 Supplier Medchemexpress LLC Supplier No. HY13970750MG
Thalidomide-NH-CBP/p300 ligand 2 (P-007) is a PROTAC-based CBP and p300 degrader. It is for research use only and not sold to patients. This compound is extracted from patent WO2020173440 and targets Cereblon.
  • PROTAC-based degrader
  • Targets CBP and p300
  • For research use only