Learn More
Medchemexpress LLC Thalidomide-NH-CBP/p300 ligand 2 | 2484739-21-9 | 98.6% | C48H57F2N11O6 | 10MG
SDP
Catalog No. 5000258562
Encompass_Preferred
Thalidomide-NH-CBP/p300 ligand 2 is a thalidomide-based PROTAC small molecule designed to induce proteasomal degradation of the transcriptional coactivators CBP and p300. It couples a thalidomide-derived E3 ligase recruiting moiety to a CBP/p300-binding ligand and is provided as a light yellow solid for research use.
  • Induces proteasomal degradation of CBP and p300.
  • Employs a thalidomide-derived E3 ligase recruiting moiety.
  • Designed for use in cellular and mechanistic research applications.
  • High purity suitable for research (reported ~98.6%).
  • Provided as a light yellow solid with molecular weight ~922.03 g·mol⁻¹.
Catalog No. 50-002-58562 Supplier Medchemexpress LLC Supplier No. HY13970710MG
Thalidomide-NH-CBP/p300 ligand 2 is a thalidomide-based PROTAC small molecule designed to induce proteasomal degradation of the transcriptional coactivators CBP and p300. It couples a thalidomide-derived E3 ligase recruiting moiety to a CBP/p300-binding ligand and is provided as a light yellow solid for research use.
  • Induces proteasomal degradation of CBP and p300.
  • Employs a thalidomide-derived E3 ligase recruiting moiety.
  • Designed for use in cellular and mechanistic research applications.
  • High purity suitable for research (reported ~98.6%).
  • Provided as a light yellow solid with molecular weight ~922.03 g·mol⁻¹.