Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-NH-CBP/p300 ligand 2 | 2484739-21-9 | MFCD28156545; MFCD34602757 | 98.6% | 922.03 g·mol⁻¹ | C48H57F2N11O6 | 5 MG
SDP

Catalog No. 5000221166
Encompass_Preferred

Thalidomide-NH-CBP/p300 ligand 2 is a PROTAC-based degrader designed to induce degradation of the CBP and p300 acetyltransferases for research applications. The compound is provided as a characterized solid with reported physicochemical and storage data for use in targeted protein degradation studies and assay development.

  • High purity (≈98.6%).
  • Molecular weight 922.03 g·mol⁻¹.
  • Chemical formula C48H57F2N11O6.
  • Soluble in DMSO at high concentration; may require sonication.
  • Solid form, light yellow to green yellow in color.
  • Recommended storage sealed, away from moisture and light.
  • Intended for research use only.

Catalog No. 50-002-21166 Supplier Medchemexpress LLC Supplier No. HY1397075MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-NH-CBP/p300 ligand 2 is a PROTAC-based degrader designed to induce degradation of the CBP and p300 acetyltransferases for research applications. The compound is provided as a characterized solid with reported physicochemical and storage data for use in targeted protein degradation studies and assay development.

  • High purity (≈98.6%).
  • Molecular weight 922.03 g·mol⁻¹.
  • Chemical formula C48H57F2N11O6.
  • Soluble in DMSO at high concentration; may require sonication.
  • Solid form, light yellow to green yellow in color.
  • Recommended storage sealed, away from moisture and light.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.