Learn More
Medchemexpress LLC Thalidomide-NH-CBP/p300 ligand 2 (P-007) | 2484739-21-9 | 98.6% | 922.03 | 100 MG
Supplier: Medchemexpress LLC HY139707100MG
Thalidomide-NH-CBP/p300 ligand 2 (P-007) is a PROTAC-based CBP and p300 degrader, extracted from patent WO2020173440. This product is for research use only.
- PROTAC-based CBP and p300 degrader
- Extracted from patent WO2020173440
- For research use only
- Targets cereblon