Learn More
Medchemexpress LLC Thalidomide-NH-PEG1-NH2 hydrochloride | 2154342-56-8 | 99.2% | C17H21ClN4O5 | 10MG
SDP
Catalog No. 5000261146
Encompass_Preferred
Thalidomide-NH-PEG1-NH2 hydrochloride is a thalidomide-derived cereblon E3 ligase ligand-linker conjugate supplied as the hydrochloride salt and intended as a building block for PROTAC design and small-molecule degradation strategies. It couples a cereblon-recruiting moiety with a short PEG1 diamine linker to facilitate conjugation to target-binding ligands and streamline synthesis workflows.
  • Provides thalidomide-based cereblon recruiting moiety.
  • Includes a short PEG1 diamine linker for flexibility in conjugation.
  • Supplied as hydrochloride salt for improved stability and handling.
  • High purity suitable for synthetic applications (99.2%).
  • Available in small milligram quantities for early-stage research.
Catalog No. 50-002-61146 Supplier Medchemexpress LLC Supplier No. HY13186710MG
Thalidomide-NH-PEG1-NH2 hydrochloride is a thalidomide-derived cereblon E3 ligase ligand-linker conjugate supplied as the hydrochloride salt and intended as a building block for PROTAC design and small-molecule degradation strategies. It couples a cereblon-recruiting moiety with a short PEG1 diamine linker to facilitate conjugation to target-binding ligands and streamline synthesis workflows.
  • Provides thalidomide-based cereblon recruiting moiety.
  • Includes a short PEG1 diamine linker for flexibility in conjugation.
  • Supplied as hydrochloride salt for improved stability and handling.
  • High purity suitable for synthetic applications (99.2%).
  • Available in small milligram quantities for early-stage research.