Learn More
Medchemexpress LLC Thalidomide-NH-PEG1-NH2 hydrochloride | 2154342-56-8 | MFCD32903278 | >98.0% | 368.81 | C16H21ClN4O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1318675MG
Encompass_Preferred
Thalidomide-NH-PEG1-NH2 hydrochloride is a thalidomide-based cereblon ligand-linker conjugate provided as the hydrochloride salt for use in PROTAC development. It features a short PEG linker and a terminal primary amine for conjugation, and is supplied as a lyophilized solid with recommended cold storage for stability.
  • Potent, cell-permeable PROTAC linker.
  • Contains a PEG1 linker with a terminal primary amine for conjugation.
  • Functions as a cereblon ligand for E3 ligase recruitment.
  • Provided as a lyophilized hydrochloride salt for stability.
  • High purity (>98%) suitable for research applications.
  • Store powder at -20 °C; store solutions at -80 °C under inert atmosphere.
Catalog No. 50-002-62132