Learn More
Medchemexpress LLC HY-130853 250mg Medchemexpress, Thalidomide-NH-PEG2-C2-NH-Boc CAS:2097509-40-3 Purity:>98%
SDP
Catalog No. 502030240
Encompass_Preferred
Medchemexpress, HY-130853 250mg Thalidomide-NH-PEG2-C2-NH-Boc CAS:2097509-40-3 Thalidomide-NH-PEG2-C2-NH-Boc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a PEG linker used for dBRD9 (compound 6) synthesis. dBRD9 is a selective BRD9 probe PROTAC degrader for the study of BAF complex biology.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0240 Supplier Medchemexpress LLC Supplier No. HY130853250MG
Medchemexpress, HY-130853 250mg Thalidomide-NH-PEG2-C2-NH-Boc CAS:2097509-40-3 Thalidomide-NH-PEG2-C2-NH-Boc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a PEG linker used for dBRD9 (compound 6) synthesis. dBRD9 is a selective BRD9 probe PROTAC degrader for the study of BAF complex biology.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.