Learn More
Medchemexpress LLC Thalidomide-NH-PEG2-COOH | 2412056-45-0 | 99.0% | 433.41 | 25 MG
SDP
Catalog No. 5000350125
Encompass_Preferred
Thalidomide-NH-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a linker that is utilized in PROTAC technology. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Utilizes PROTAC technology
  • Incorporates Thalidomide based cereblon ligand
  • Intended for research use only
Catalog No. 50-003-50125 Supplier Medchemexpress LLC Supplier No. 25MGHY138772
Thalidomide-NH-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a linker that is utilized in PROTAC technology. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Utilizes PROTAC technology
  • Incorporates Thalidomide based cereblon ligand
  • Intended for research use only