Learn More
Medchemexpress LLC Thalidomide-NH-PEG2-COOH | 2412056-45-0 | MFCD34368531 | ≥95.0% | 433.41 g/mol | C20H23N3O8 | 5 MG
SDP
Catalog No. 5000223317
Encompass_Preferred
Thalidomide-NH-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon (CRBN) ligand with a short PEG2 carboxylic acid linker. It is designed as a building block for PROTAC (proteolysis targeting chimera) synthesis and related chemical biology research. For research use only.
  • Provides CRBN engagement via a thalidomide-derived ligand.
  • Contains a PEG2 spacer terminating in a carboxylic acid for linker conjugation.
  • Suitable as a building block for PROTAC synthesis and E3 ligase recruitment studies.
  • Characterized by analytical data, including HPLC, LC-MS, and NMR files.
  • Available in small research-scale quantities for medicinal chemistry applications.
Catalog No. 50-002-23317 Supplier Medchemexpress LLC Supplier No. HY1387725MG
Thalidomide-NH-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon (CRBN) ligand with a short PEG2 carboxylic acid linker. It is designed as a building block for PROTAC (proteolysis targeting chimera) synthesis and related chemical biology research. For research use only.
  • Provides CRBN engagement via a thalidomide-derived ligand.
  • Contains a PEG2 spacer terminating in a carboxylic acid for linker conjugation.
  • Suitable as a building block for PROTAC synthesis and E3 ligase recruitment studies.
  • Characterized by analytical data, including HPLC, LC-MS, and NMR files.
  • Available in small research-scale quantities for medicinal chemistry applications.