Learn More
Medchemexpress LLC Thalidomide-NH-PEG2-COOH | 2412056-45-0 | MFCD34368531 | 99.0% | C20H23N3O8 | 10MG
SDP
Catalog No. 5000260682
Encompass_Preferred
Thalidomide-NH-PEG2-COOH is a thalidomide-derived cereblon (CRBN) ligand-linker conjugate designed as a building block for PROTAC and targeted protein degradation research. It couples a CRBN-recruiting thalidomide moiety to a short PEG2 spacer terminated with a carboxylic acid, facilitating conjugation to warheads or scaffolds for degrader design.
  • Thalidomide-based cereblon ligand for CRBN recruitment.
  • PEG2 linker terminated with carboxylic acid for conjugation.
  • Suitable as a building block for PROTAC synthesis and linker optimization.
  • High purity (99.0%) supporting reproducible conjugation chemistry.
  • Molecular formula C20H23N3O8; molecular weight 433.41 g/mol.
  • Supplied in small milligram pack sizes for research use.
Catalog No. 50-002-60682 Supplier Medchemexpress LLC Supplier No. HY13877210MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thalidomide-NH-PEG2-COOH is a thalidomide-derived cereblon (CRBN) ligand-linker conjugate designed as a building block for PROTAC and targeted protein degradation research. It couples a CRBN-recruiting thalidomide moiety to a short PEG2 spacer terminated with a carboxylic acid, facilitating conjugation to warheads or scaffolds for degrader design.
  • Thalidomide-based cereblon ligand for CRBN recruitment.
  • PEG2 linker terminated with carboxylic acid for conjugation.
  • Suitable as a building block for PROTAC synthesis and linker optimization.
  • High purity (99.0%) supporting reproducible conjugation chemistry.
  • Molecular formula C20H23N3O8; molecular weight 433.41 g/mol.
  • Supplied in small milligram pack sizes for research use.