Learn More
Medchemexpress LLC Thalidomide-NH-PEG3-COOH | 2375283-62-6 | 98.9% | 477.46 | 100 MG
SDP
Catalog No. 5000352083
Encompass_Preferred
Thalidomide-NH-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a thalidomide-based cereblon ligand and a linker utilized in PROTAC technology. This product is intended for research purposes only.
  • Incorporates a thalidomide-based cereblon ligand
  • Designed for use in PROTAC technology
  • Targets cereblon
  • Appears as a white to yellow solid
  • Useful in cancer research applications
  • Functions as an E3 ligase ligand-linker conjugate
  • Involved in autophagy and apoptosis studies
  • Acts as an inhibitor
Catalog No. 50-003-52083 Supplier Medchemexpress LLC Supplier No. HY138771100MG
Thalidomide-NH-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a thalidomide-based cereblon ligand and a linker utilized in PROTAC technology. This product is intended for research purposes only.
  • Incorporates a thalidomide-based cereblon ligand
  • Designed for use in PROTAC technology
  • Targets cereblon
  • Appears as a white to yellow solid
  • Useful in cancer research applications
  • Functions as an E3 ligase ligand-linker conjugate
  • Involved in autophagy and apoptosis studies
  • Acts as an inhibitor