Learn More
Medchemexpress LLC Thalidomide-NH-PEG3-COOH | 2375283-62-6 | 98.9% | 477.46 | C22H27N3O9 | 5 MG
SDP
Catalog No. 5000223316
Encompass_Preferred
Thalidomide-NH-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a three-unit polyethylene glycol (PEG3) linker terminating in a carboxylic acid. It is intended as a modular building block for PROTAC development to recruit CRBN and enable linker attachment to target-binding ligands.
  • Designed for PROTAC assembly and CRBN recruitment.
  • Contains a PEG3 linker terminating in a carboxylic acid for facile conjugation.
  • Incorporates a thalidomide-based cereblon ligand.
  • High chemical purity (≈98.9%).
  • Suitable for medicinal chemistry and chemical biology applications.
Catalog No. 50-002-23316 Supplier Medchemexpress LLC Supplier No. HY1387715MG
Thalidomide-NH-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a three-unit polyethylene glycol (PEG3) linker terminating in a carboxylic acid. It is intended as a modular building block for PROTAC development to recruit CRBN and enable linker attachment to target-binding ligands.
  • Designed for PROTAC assembly and CRBN recruitment.
  • Contains a PEG3 linker terminating in a carboxylic acid for facile conjugation.
  • Incorporates a thalidomide-based cereblon ligand.
  • High chemical purity (≈98.9%).
  • Suitable for medicinal chemistry and chemical biology applications.