Learn More
Medchemexpress LLC Thalidomide-NH-PEG3-COOH | 2375283-62-6 | 98.9% | C22H27N3O9 | 10MG
SDP
Catalog No. 5000262031
Encompass_Preferred
Thalidomide-NH-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate that links a thalidomide-derived cereblon (CRBN) ligand to a three-unit polyethylene glycol (PEG3) spacer terminated with a carboxylic acid. It is designed as a building block for PROTAC synthesis to recruit the CRBN E3 ligase and enable conjugation to target-binding ligands.
  • High reported purity (98.9%).
  • Includes a thalidomide-derived cereblon ligand for CRBN recruitment.
  • PEG3 spacer provides flexible, hydrophilic linkage.
  • Terminal carboxylic acid enables standard coupling chemistries.
  • Soluble in DMSO for common assay and synthesis workflows.
  • Supplied as a solid suitable for storage and handling in research labs.
Catalog No. 50-002-62031 Supplier Medchemexpress LLC Supplier No. HY13877110MG
Thalidomide-NH-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate that links a thalidomide-derived cereblon (CRBN) ligand to a three-unit polyethylene glycol (PEG3) spacer terminated with a carboxylic acid. It is designed as a building block for PROTAC synthesis to recruit the CRBN E3 ligase and enable conjugation to target-binding ligands.
  • High reported purity (98.9%).
  • Includes a thalidomide-derived cereblon ligand for CRBN recruitment.
  • PEG3 spacer provides flexible, hydrophilic linkage.
  • Terminal carboxylic acid enables standard coupling chemistries.
  • Soluble in DMSO for common assay and synthesis workflows.
  • Supplied as a solid suitable for storage and handling in research labs.