Learn More
Medchemexpress LLC Thalidomide-NH-PEG3-NH-Boc | 2204246-03-5 | 97.2% | 548.59 | 100 MG

Supplier: Medchemexpress LLC HY44063100MG
Thalidomide-NH-PEG3-NH-Boc is a chemical compound with a molecular weight of 548.59. It appears as a light yellow to yellow oil and its structure is consistent with LCMS analysis. This product has been tested and complies with the given specifications.
- Molecular formula: C26H36N4O9
- Storage in pure form: -20°C for 3 years; 4°C for 2 years
- Storage in solvent: -80°C for 6 months; -20°C for 1 month
- Purity: 97.20%
- Appearance: Light yellow to yellow (oil)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.