Learn More
Medchemexpress LLC Thalidomide-NH-PEG3-propionic acid | 2138440-82-9 | 99.7% | 477.46 | 100 MG
SDP
Catalog No. 5000364862
Encompass_Preferred
Thalidomide-NH-PEG3-propionic acid is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a 3-unit PEG linker used in PROTAC technology. This product is for research use only, intended for experienced personnel in appropriately equipped and authorized facilities.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Uses a 3-unit PEG linker
  • Utilized in PROTAC technology
  • Exploits intracellular ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-64862 Supplier Medchemexpress LLC Supplier No. 100MGHY136166
Thalidomide-NH-PEG3-propionic acid is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a 3-unit PEG linker used in PROTAC technology. This product is for research use only, intended for experienced personnel in appropriately equipped and authorized facilities.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Uses a 3-unit PEG linker
  • Utilized in PROTAC technology
  • Exploits intracellular ubiquitin-proteasome system for selective protein degradation