Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-NH-PEG4-COOH | 2412056-48-3 | 96.7% | 521.52 | 50 MG
SDP

Catalog No. 5000365992
Encompass_Preferred

Thalidomide-NH-PEG4-COOH is an E3 ligase ligand-linker conjugate which can be used for synthesizing dCBP-1. dCBP-1 is a potent and selective heterobifunctional degrader of p300/CBP.

  • E3 ligase ligand-linker conjugate
  • Used for synthesizing dCBP-1
  • Potent and selective heterobifunctional degrader of p300/CBP
  • Purity of 96.7%
  • Appearance: yellow to green (solid-liquid mixture)
  • Store at -20°C, protect from light

Catalog No. 50-003-65992 Supplier Medchemexpress LLC Supplier No. 50MGHY134591
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-NH-PEG4-COOH is an E3 ligase ligand-linker conjugate which can be used for synthesizing dCBP-1. dCBP-1 is a potent and selective heterobifunctional degrader of p300/CBP.

  • E3 ligase ligand-linker conjugate
  • Used for synthesizing dCBP-1
  • Potent and selective heterobifunctional degrader of p300/CBP
  • Purity of 96.7%
  • Appearance: yellow to green (solid-liquid mixture)
  • Store at -20°C, protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.