Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide O-amido C3-COOH | 2308035-51-8 | 98.4% | 417.37 g/mol | C19H19N3O8 | 50 MG
SDP

Catalog No. 5000247795
Encompass_Preferred

Thalidomide O-amido C3-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon-binding moiety with a short linker terminating in a carboxylic acid. It is provided as a research-grade chemical for use in PROTAC design and related chemical biology applications.

  • Contains thalidomide-based cereblon ligand
  • Three-carbon (C3) linker with terminal carboxylic acid
  • Molecular formula C19H19N3O8; molecular weight 417.37 g/mol
  • Purity 98.4% (research grade)
  • Supplied as a white solid suitable for chemical synthesis
  • Intended for research use only, not for human or clinical use

Catalog No. 50-002-47795 Supplier Medchemexpress LLC Supplier No. HY10361250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide O-amido C3-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon-binding moiety with a short linker terminating in a carboxylic acid. It is provided as a research-grade chemical for use in PROTAC design and related chemical biology applications.

  • Contains thalidomide-based cereblon ligand
  • Three-carbon (C3) linker with terminal carboxylic acid
  • Molecular formula C19H19N3O8; molecular weight 417.37 g/mol
  • Purity 98.4% (research grade)
  • Supplied as a white solid suitable for chemical synthesis
  • Intended for research use only, not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.