Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-o-amido-c3-cooh | 2308035-51-8 | MFCD31560483 | >95.0% | 417.4 g/mol | C19H19N3O8 | 500 MG
SDP

Supplier:  Medchemexpress LLC HY103612500MG

Encompass_Preferred

Thalidomide-O-amido-C3-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon (CRBN) ligand with a three-carbon linker terminating in a carboxylic acid. It is intended for use in PROTAC synthesis and related chemical biology research, enabling conjugation to target-binding ligands for targeted protein degradation studies.

  • Synthesized E3 ligase ligand-linker conjugate suitable for PROTAC design.
  • Contains a thalidomide-derived cereblon ligand for CRBN recruitment.
  • Features a C3 linker terminating in a carboxylic acid for conjugation.
  • Available as a solid research-grade reagent in multiple package sizes.
  • Recommended storage: 4°C under inert gas; in solvent store at -80°C for long term.
  • Hazard: handle with appropriate personal protective equipment; may cause skin and eye irritation and is harmful if swallowed.

Catalog No. 50-002-44964


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.